Assay Details
Target Gene Details
Entrez Gene ID: | 20822 |
Gene Name: | TROVE domain family, member 2 |
Gene Aliases: |
1810007I17Rik, A530054J02Rik, AI646302, SS-A/Ro, Ssa, Ssa2 |
Location: |
Chr.1:143750790-143777051 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Trove2 | NM_013835.2 | 9 | 5703 | NP_038863.1 |
AK040977.1 | 1 | 901 | ||
AK078995.1 | 1 | 409 | ||
AK079759.1 | 1 | 885 |
More Information
Set Membership: |
Intragenic Exonic |