Assay Details
Target Gene Details
Entrez Gene ID: | 433091 |
Gene Name: | patatin-like phospholipase domain containing 1 |
Gene Aliases: |
- |
Location: |
Chr.17:28857965-28890308 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pnpla1 | NM_001034885.3 | 10 | 3409 | NP_001030057.1 |
AK132521.1 | 10 | 3410 | BAE21216.1 | |
BC139043.1 | 10 | 3116 |
More Information
Set Membership: |
Intragenic Exonic |