Assay Details
Target Gene Details
Entrez Gene ID: | 12892 |
Gene Name: | coproporphyrinogen oxidase |
Gene Aliases: |
CPX, Cpo, HCP, cac, nct |
Location: |
Chr.16:58670208-58680391 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cpox | NM_007757.2 | 7 | 1727 | NP_031783.2 |
AK166512.1 | 7 | 1590 | ||
BC017680.1 | 7 | 1584 | ||
D16333.1 | 7 | 1641 | BAA03840.1 |
More Information
Set Membership: |
Intragenic Exonic |