Assay Details
Target Gene Details
Entrez Gene ID: | 17472 |
Gene Name: | guanylate binding protein 4 |
Gene Aliases: |
AW228052, Mag-2, Mpa-2, Mpa2, mKIAA4245 |
Location: |
Chr.5:105115767-105139630 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gbp4 | NM_001256005.1 | 11 | 3117 | NP_001242934.1 |
NM_008620.3 | 11 | 3054 | NP_032646.2 | |
AA177993.1 | 1 | 97 | ||
AK220567.1 | 11 | 3035 | BAD90331.1 | |
EF494423.1 | 11 | 3117 | ||
EF494424.1 | 11 | 3088 |
More Information
Set Membership: |
Intragenic Exonic |