Assay Details
Target Gene Details
Entrez Gene ID: | 11909 |
Gene Name: | activating transcription factor 2 |
Gene Aliases: |
Atf-2, CRE-BP, Creb2, D130078H02Rik, D18875, Tg(Gzma-Klra1)7Wum, mXBP |
Location: |
Chr.2:73816509-73892639 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Atf2 | NM_001025093.2 | 15 | 3947 | NP_001020264.1 |
NM_001284369.1 | 13 | 3787 | NP_001271298.1 | |
NM_001284372.1 | 13 | 3653 | NP_001271301.1 | |
NM_001284373.1 | 12 | 3590 | NP_001271302.1 | |
NM_001284374.1 | 12 | 3672 | NP_001271303.1 | |
NM_001284376.1 | 15 | 3829 | NP_001271305.1 | |
NM_009715.3 | 14 | 3850 | NP_033845.1 | |
AK144977.1 | 13 | 3627 | ||
AK145887.1 | 14 | 3229 | BAE26726.1 | |
BC042210.1 | 12 | 3623 | AAH42210.1 | |
BC079883.1 | 14 | 3772 | AAH79883.1 | |
BC082596.1 | 12 | 3536 | AAH82596.1 |
More Information
Set Membership: |
Intragenic Exonic |