Assay Details
Target Gene Details
Entrez Gene ID: | 50500 |
Gene Name: | tocopherol (alpha) transfer protein |
Gene Aliases: |
alpha-TTP |
Location: |
Chr.4:20008089-20030785 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ttpa | NM_015767.3 | 5 | 2898 | NP_056582.1 |
AK050374.1 | 5 | 2899 | BAC34218.1 |
More Information
Set Membership: |
Intragenic Exonic |