Assay Details
Target Gene Details
Entrez Gene ID: | 65103 |
Gene Name: | ADP-ribosylation factor-like 6 interacting protein 6 |
Gene Aliases: |
2310057C01Rik, 2610529A11Rik, Aip-6 |
Location: |
Chr.2:53192084-53219221 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Arl6ip6 | NM_022989.4 | 4 | 773 | NP_075365.3 |
AK011070.1 | 4 | 727 | BAB27377.1 | |
AK016267.1 | 4 | 748 | BAB30169.1 | |
AK032740.1 | 4 | 747 | BAC28004.1 | |
AK035521.1 | 4 | 770 | BAC29090.1 | |
AK052010.1 | 4 | 747 | BAC34831.1 | |
AK082824.1 | 4 | 745 | BAC38638.1 | |
AK159990.1 | 4 | 735 | BAE35540.1 | |
BC019550.1 | 4 | 740 | AAH19550.1 | |
BC157980.1 | 4 | 764 |