Assay Details
Target Gene Details
Entrez Gene ID: | 70465 |
Gene Name: | WD repeat domain 77 |
Gene Aliases: |
2610003I18Rik, 2610312E17Rik, C79984, p44/MEP50 |
Location: |
Chr.3:105959498-105969760 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Wdr77 | NM_027432.3 | 10 | 1176 | NP_081708.1 |
AK032897.1 | 8 | 715 | BAC28076.1 | |
AK048488.1 | 10 | 1199 | BAC33350.1 | |
AK050391.1 | 10 | 1176 | BAC34232.1 | |
AK078856.1 | 11 | 1135 | ||
AK131608.1 | 9 | 1087 | BAE20717.1 | |
AK161812.1 | 9 | 1634 | ||
AK168006.1 | 10 | 1178 | ||
AK168752.1 | 10 | 1198 | ||
AK169128.1 | 10 | 1177 | BAE40907.1 | |
AK214407.1 | 1 | 180 | ||
BC005755.1 | 10 | 1131 | AAH05755.1 |