Assay Details
Target Gene Details
Entrez Gene ID: | 17187 |
Gene Name: | Max protein |
Gene Aliases: |
AA960152, AI875693, bHLHd4, bHLHd5, bHLHd6, bHLHd7, bHLHd8 |
Location: |
Chr.12:76937269-76962335 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Max | NM_001146176.1 | 4 | 1677 | NP_001139648.1 |
NM_008558.2 | 5 | 1704 | NP_032584.2 | |
AK080431.1 | 4 | 1632 | BAC37914.1 | |
AY423852.1 | ||||
M63903.1 | 5 | 1641 | AAA39797.1 |
More Information
Set Membership: |
Intragenic Exonic |