Assay Details
Target Gene Details
Entrez Gene ID: | 83602 |
Gene Name: | general transcription factor II A, 1 |
Gene Aliases: |
6330549H03Rik, AA536742, AA959775, AW060250, TfIIAa/b, Tfiia1 |
Location: |
Chr.12:91555262-91590487 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gtf2a1 | NM_031391.2 | 9 | 5283 | NP_113568.2 |
NM_175335.3 | 9 | 5270 | NP_780544.1 | |
AL389955.1 | 1 | 443 | ||
AW552430.1 | 1 | 421 | ||
BX526091.1 |
More Information
Set Membership: |
Intragenic Exonic |