Assay Details
Target Gene Details
Entrez Gene ID: | 66816 |
Gene Name: | THAP domain containing, apoptosis associated protein 2 |
Gene Aliases: |
2900040O07Rik, 9030625G08Rik, AI450385, AI649097 |
Location: |
Chr.10:115369966-115384435 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Thap2 | NM_025780.3 | 3 | 512 | NP_080056.1 |
AK009162.1 | 3 | 491 | ||
AK018577.1 | 3 | 492 | BAB31288.1 | |
AK051198.1 | 2 | 292 | ||
AK081303.1 | 3 | 513 | BAC38189.1 | |
BC027502.1 | 3 | 420 | AAH27502.1 |