Assay Details
Target Gene Details
Entrez Gene ID: | 14874 |
Gene Name: | glutathione transferase zeta 1 (maleylacetoacetate isomerase) |
Gene Aliases: |
MAAI |
Location: |
Chr.12:87147165-87164723 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gstz1 | NM_001252555.1 | 10 | 1049 | NP_001239484.1 |
NM_001252556.1 | 8 | 678 | NP_001239485.1 | |
NM_010363.4 | 9 | 746 | NP_034493.1 | |
AB041613.1 | 10 | 1049 | BAA95096.1 | |
AF093418.1 | 9 | 695 | AAD43846.1 | |
AK002398.1 | 9 | 703 | BAB22070.1 | |
AK012796.1 | 9 | 747 | ||
AK075927.1 | 9 | 700 | BAC36059.1 | |
BC031777.1 | 9 | 735 | AAH31777.1 |
More Information
Set Membership: |
Intragenic Exonic |