Assay Details
Target Gene Details
Entrez Gene ID: | 23942 |
Gene Name: | metastasis-associated gene family, member 2 |
Gene Aliases: |
AW550797, Mata1l1, Mta1l1, mmta2 |
Location: |
Chr.19:8941920-8952300 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mta2 | NM_011842.3 | 1 | 387 | NP_035972.3 |
AF159259.1 | 1 | 262 | AAD51281.1 | |
AK077548.1 | 1 | 369 | ||
AK088158.1 | 1 | 300 | BAC40180.1 | |
AK147099.1 | 1 | 366 | BAE27675.1 | |
AK149834.1 | 1 | 433 | BAE29113.1 | |
AK157717.1 | 1 | 291 | BAE34165.1 | |
AK159979.1 | 1 | 363 | BAE35530.1 | |
BC079847.1 | 1 | 387 | AAH79847.1 |
More Information
Set Membership: |
Intragenic Exonic |