Assay Details
Target Gene Details
Entrez Gene ID: | 70942 |
Gene Name: | RIKEN cDNA 4931403E22 gene |
Gene Aliases: |
- |
Location: |
Chr.19:26768801-26823907 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
4931403E22Rik | NR_045306.1 | |||
AK016426.1 | ||||
BY716074.1 |
Target Gene Details
Entrez Gene ID: | 67155 |
Gene Name: | SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily a, member 2 |
Gene Aliases: |
2610209L14Rik, SNF2alpha, Snf2l2, brahma, brm |
Location: |
Chr.19:26605089-26778321 on Build GRCm38 |
Assay Gene Location: | Within Exon 36 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Smarca2 | NM_011416.2 | 33 | 5645 | NP_035546.2 |
NM_026003.2 | 9 | 1794 | NP_080279.1 | |
AI842968.1 | 1 | 185 | ||
AK009309.1 | 5 | 1304 | ||
AK011935.1 | 9 | 1739 | BAB27925.1 | |
AK147322.1 | 33 | 5794 | BAE27845.1 | |
BC004653.1 | 5 | 1292 | AAH04653.1 | |
BC022975.1 | 11 | 2271 | AAH22975.1 | |
BC056199.1 | 23 | 3743 | AAH56199.1 | |
BC075641.1 | 33 | 5627 | AAH75641.1 |
More Information
Set Membership: |
Intragenic Exonic Intronic Non-exonic |