Assay Details
Target Gene Details
Entrez Gene ID: | 23963 |
Gene Name: | teneurin transmembrane protein 1 |
Gene Aliases: |
Odz1, Odz2, TCAP-1, Ten-m1, ten-1 |
Location: |
Chr.X:42527866-43429156 on Build GRCm38 |
Assay Gene Location: | Within Exon 29 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tenm1 | NM_011855.4 | 29 | 5579 | NP_035985.2 |
AB025410.1 | 29 | 5581 | BAA77396.1 | |
AK037897.1 | 2 | 268 | BAC29893.1 | |
BC138860.1 | 28 | 5562 | ||
BC138861.1 | 28 | 5562 |