Assay Details
Target Gene Details
Entrez Gene ID: | 109113 |
Gene Name: | ubiquitin-like, containing PHD and RING finger domains 2 |
Gene Aliases: |
2310065A22Rik, AI426270, AW214556, D130071B19Rik, Nirf |
Location: |
Chr.19:30030433-30093729 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Uhrf2 | NM_144873.2 | 3 | 737 | NP_659122.2 |
AB116653.1 | 3 | 472 | BAC81739.1 | |
AF274047.1 | 3 | 647 | AAM33798.1 | |
AK031036.1 | 3 | 799 | BAC27224.1 | |
AK041564.1 | 3 | 759 | BAC30987.1 | |
AK042321.1 | 3 | 778 | BAC31223.1 | |
AK051743.1 | 3 | 778 | BAC34750.1 | |
AK080925.1 | 3 | 778 | BAC38081.1 | |
AK082518.1 | ||||
AK172682.1 | 3 | 621 | BAE43129.1 | |
AK184966.1 | ||||
AK206406.1 | ||||
BC060241.1 | 3 | 737 | AAH60241.1 |
More Information
Set Membership: |
Intragenic Exonic |