Assay Details
Target Gene Details
Entrez Gene ID: | 58205 |
Gene Name: | programmed cell death 1 ligand 2 |
Gene Aliases: |
B7-DC, Btdc, F730015O22Rik, PD-L2 |
Location: |
Chr.19:29406240-29472927 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pdcd1lg2 | NM_021396.2 | 3 | 541 | NP_067371.1 |
AF142780.2 | 3 | 514 | AAD33892.1 | |
AK089369.1 | 3 | 544 | BAC40858.1 | |
AK141111.1 | 2 | 631 | BAE24564.1 | |
AK155002.1 | 3 | 481 | BAE32986.1 | |
BC104138.1 | 3 | 446 | AAI04139.1 | |
BC104139.1 | 3 | 446 | AAI04140.1 | |
BY749628.1 | 3 | 544 |
More Information
Set Membership: |
Intragenic Exonic |