Assay Details
Target Gene Details
Entrez Gene ID: | 70044 |
Gene Name: | terminal uridylyl transferase 1, U6 snRNA-specific |
Gene Aliases: |
2700038E08Rik, PAPD2, Rbm21, TUTase6 |
Location: |
Chr.19:8953850-8966210 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tut1 | NM_197993.2 | 3 | 409 | NP_932110.1 |
AK138418.1 | 3 | 329 | BAE23652.1 | |
AK179054.1 | ||||
BC023900.1 | 3 | 386 | AAH23900.1 | |
BC025499.1 | 3 | 433 | AAH25499.1 |
More Information
Set Membership: |
Intragenic Exonic |