Assay Details
Target Gene Details
Entrez Gene ID: | 71738 |
Gene Name: | MAM domain containing 2 |
Gene Aliases: |
1200015L10Rik, AI591564, mamcan |
Location: |
Chr.19:23302609-23448500 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mamdc2 | NM_174857.3 | 9 | 1624 | NP_777282.1 |
AK004794.1 | 8 | 1487 | ||
AK035566.1 | 9 | 1625 | BAC29108.1 | |
AK154219.1 | 9 | 1701 | BAE32442.1 | |
BC042773.1 | 9 | 1653 | AAH42773.1 |