Assay Details
Target Gene Details
Entrez Gene ID: | 56248 |
Gene Name: | adenylate kinase 3 |
Gene Aliases: |
AA407498, AI506714, AK-3, Ak3l, Ak3l1, Akl3l |
Location: |
Chr.19:28995380-29048672 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ak3 | NM_021299.1 | 5 | 998 | NP_067274.1 |
AB020203.1 | 5 | 901 | BAA77360.1 | |
AK004864.2 | 5 | 1013 | BAB23625.1 | |
AK005194.2 | 5 | 1052 | BAB23876.1 | |
AK008681.1 | 5 | 1015 | BAB25829.1 | |
AK031631.1 | 5 | 996 | BAC27488.1 | |
AK053644.1 | 5 | 1058 | BAC35459.1 | |
AK089027.1 | 5 | 1049 | BAC40707.1 | |
AK147060.1 | 5 | 1058 | BAE27644.1 | |
AK149996.1 | 5 | 1055 | BAE29224.1 | |
AK195026.1 | ||||
AK216638.1 | ||||
BC016432.1 | 5 | 1000 | AAH16432.1 | |
BC019174.1 | 5 | 1051 | AAH19174.1 | |
BC024871.1 | 5 | 998 | AAH24871.1 | |
BC058191.1 | 5 | 1028 | AAH58191.1 |
More Information
Set Membership: |
Intragenic Exonic |