Assay Details
Target Gene Details
Entrez Gene ID: | 209268 |
Gene Name: | immunoglobulin superfamily, member 1 |
Gene Aliases: |
5330413N23, 5530402E03, AI747649, InhBP/p120, mKIAA0364 |
Location: |
Chr.X:49782536-49797906 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Igsf1 | NM_177591.4 | 8 | 1480 | NP_808259.2 |
NM_183335.2 | 7 | 1344 | NP_899178.2 | |
AK129126.1 | 8 | 1462 | BAC97936.1 | |
AY227771.1 | 8 | 1474 | AAP57079.1 | |
AY227773.1 | 7 | 1344 | AAP57081.1 |
More Information
Set Membership: |
Intragenic Exonic |