Assay Details
Target Gene Details
Entrez Gene ID: | 225875 |
Gene Name: | leucine rich repeat and fibronectin type III domain containing 4 |
Gene Aliases: |
BC023156, Salm3 |
Location: |
Chr.19:4611784-4615667 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Lrfn4 | NM_153388.4 | 2 | 931 | NP_700437.2 |
AK081560.1 | 1 | 1215 | BAC38259.1 | |
AK163603.1 | 2 | 789 | BAE37415.1 | |
BC023036.1 | 1 | 544 | AAH23036.1 | |
BC023156.1 | 2 | 587 | AAH23156.1 | |
DQ078786.1 | 1 | 216 | AAZ23615.1 |
Target Gene Details
Entrez Gene ID: | 18563 |
Gene Name: | pyruvate carboxylase |
Gene Aliases: |
Pc, Pcb |
Location: |
Chr.19:4510472-4621752 on Build GRCm38 |
Assay Gene Location: | Within Intron 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pcx | NM_001162946.1 | NP_001156418.1 | ||
NM_008797.3 | NP_032823.2 | |||
AK077665.1 | BAC36939.1 | |||
AK148328.1 | BAE28484.1 | |||
AK170596.1 | BAE41902.1 | |||
AK170871.1 | ||||
AK172320.1 | BAE42943.1 | |||
BC055030.1 | AAH55030.1 | |||
L09192.1 | AAA39737.1 | |||
M97957.1 | AAA39897.1 |
More Information
Set Membership: |
Intragenic Exonic Intronic Non-exonic |