Assay Details
Target Gene Details
Entrez Gene ID: | 81500 |
Gene Name: | endoplasmic reticulum chaperone SIL1 homolog (S. cerevisiae) |
Gene Aliases: |
1810057E01Rik, AI042831, wz |
Location: |
Chr.18:35266396-35499052 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Sil1 | NM_030749.2 | 10 | 1440 | NP_109674.2 |
AJ297884.1 | 10 | 1381 | CAC17789.1 | |
AK170415.1 | 10 | 1393 | BAE41781.1 | |
AK189955.1 | ||||
AK206841.1 | ||||
AK208966.1 | ||||
AK215511.1 | ||||
AK220123.1 | ||||
BC016119.1 | 10 | 1417 | AAH16119.1 | |
BC016466.1 | 10 | 1403 | AAH16466.1 | |
BP771607.1 | 1 | 302 | ||
BX512650.1 | 2 | 312 |