Assay Details
Target Gene Details
Entrez Gene ID: | 14544 |
Gene Name: | guanine deaminase |
Gene Aliases: |
AU015411, AW047581 |
Location: |
Chr.19:21391307-21472661 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gda | NM_010266.2 | 14 | 2127 | NP_034396.1 |
AF174583.1 | 14 | 2053 | AAD50297.1 | |
AK041056.1 | 14 | 2066 | BAC30802.1 | |
AK079419.1 | 14 | 1935 | ||
AK079459.1 | 14 | 2066 | ||
AK149199.1 | 1 | 204 | ||
AK157014.1 | 14 | 2089 | BAE33931.1 | |
AK173138.1 | 14 | 2056 | BAD32416.1 | |
BC076583.1 | AAH76583.1 |
More Information
Set Membership: |
Intragenic Exonic |