Assay Details
Target Gene Details
Entrez Gene ID: | 66990 |
Gene Name: | transmembrane protein 134 |
Gene Aliases: |
2410001H17Rik, AI463452 |
Location: |
Chr.19:4125960-4132307 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tmem134 | NM_001078649.1 | 7 | 1350 | NP_001072117.1 |
NM_025889.2 | 6 | 1305 | NP_080165.1 | |
AK009263.1 | 5 | 1427 | ||
AK009745.1 | 6 | 1281 | BAB26475.1 | |
AK010323.1 | 7 | 1325 | BAC25290.1 |