Assay Details
Target Gene Details
Entrez Gene ID: | 16601 |
Gene Name: | Kruppel-like factor 9 |
Gene Aliases: |
2310051E17Rik, AA589643, BTEB-1, Bteb1, ENSMUSG00000055440, Gm9971 |
Location: |
Chr.19:23141226-23166911 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Klf9 | NM_010638.4 | 1 | 863 | NP_034768.2 |
AK028544.1 | 1 | 864 | BAC26000.1 | |
AK208900.1 | ||||
BC138724.1 | 1 | 676 | ||
BC138725.1 | 1 | 676 | ||
CT010185.1 | 1 | 334 | CAJ18393.1 | |
Y14296.1 | 1 | 334 | CAA74671.1 |
More Information
Set Membership: |
Intragenic Exonic |