Assay Details
Target Gene Details
Entrez Gene ID: | 60533 |
Gene Name: | CD274 antigen |
Gene Aliases: |
A530045L16Rik, B7h1, Pdcd1l1, Pdcd1lg1, Pdl1 |
Location: |
Chr.19:29367438-29388095 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cd274 | NM_021893.3 | 4 | 571 | NP_068693.1 |
AF233517.1 | 3 | 504 | AAG18509.1 | |
AF317088.1 | 3 | 488 | AAG31810.1 | |
AK154403.1 | 4 | 550 | BAE32562.1 | |
BC066841.1 | 4 | 554 | AAH66841.1 | |
GQ904196.1 | 3 | 488 |
More Information
Set Membership: |
Intragenic Exonic |