Assay Details
Target Gene Details
Entrez Gene ID: | 14625 |
Gene Name: | glycerol kinase-like 1 |
Gene Aliases: |
AI449806, Gk-rs1 |
Location: |
Chr.18:52693679-52695593 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gykl1 | NM_010293.3 | 1 | 1589 | NP_034423.2 |
AF117733.1 | 1 | 1589 | AAD24550.1 | |
AK076631.1 | 1 | 1565 | BAC36422.1 | |
BC119574.2 | 1 | 1570 | ||
BY434724.1 | 1 | 328 |
More Information
Set Membership: |
Intragenic Exonic |