Assay Details
Target Gene Details
Entrez Gene ID: | 210510 |
Gene Name: | tudor domain containing 6 |
Gene Aliases: |
AB097085, Tdr2 |
Location: |
Chr.17:43615334-43631350 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tdrd6 | NM_001161366.1 | 5 | 6970 | NP_001154838.1 |
NM_001161367.1 | 5 | 6949 | NP_001154839.1 | |
NM_198418.2 | 5 | 6967 | NP_940810.2 | |
AB097085.1 | 5 | 6967 | BAD01486.1 | |
BU605232.1 | 1 | 395 |
More Information
Set Membership: |
Intragenic Exonic |