Assay Details
Target Gene Details
Entrez Gene ID: | 12393 |
Gene Name: | runt related transcription factor 2 |
Gene Aliases: |
AML3, Cbf, Cbfa-1, Cbfa1, LS3, Osf2, PEBP2aA, Pebp2a1, Pebpa2a |
Location: |
Chr.17:44495986-44814831 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Runx2 | NM_001145920.2 | 7 | 4280 | NP_001139392.1 |
NM_001146038.2 | 9 | 3545 | NP_001139510.1 | |
NM_001271627.1 | 8 | 3709 | NP_001258556.1 | |
NM_001271630.1 | 6 | 3634 | NP_001258559.1 | |
NM_001271631.1 | 5 | 3529 | NP_001258560.1 | |
NM_009820.5 | 9 | 3512 | NP_033950.2 | |
NR_073392.1 | 8 | 3296 | ||
NR_073425.1 | ||||
AK134925.1 | ||||
AK158925.1 | ||||
AK164454.1 | 1 | 178 | ||
D14636.1 | 7 | 4278 | BAA03485.1 | |
D14637.1 | BAA03486.1 |
More Information
Set Membership: |
Intragenic Exonic |