Assay Details
Target Gene Details
Entrez Gene ID: | 54419 |
Gene Name: | claudin 6 |
Gene Aliases: |
AL024037 |
Location: |
Chr.17:23679348-23682446 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cldn6 | NM_018777.4 | 3 | 1268 | NP_061247.1 |
AK010560.1 | 3 | 1269 | BAB27028.1 | |
AK010682.1 | 3 | 1263 | BAB27114.1 | |
BC005718.1 | 3 | 1245 | AAH05718.1 | |
BC050138.1 | 3 | 1251 | AAH50138.1 | |
BU757997.1 | 1 | 254 |
More Information
Set Membership: |
Intragenic Exonic |