Assay Details
Target Gene Details
Entrez Gene ID: | 50873 |
Gene Name: | Parkinson disease (autosomal recessive, juvenile) 2, parkin |
Gene Aliases: |
PRKN |
Location: |
Chr.17:10840384-12063370 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Park2 | NM_016694.3 | 12 | 2295 | NP_057903.1 |
AF250293.1 | 12 | 2295 | AAG13890.1 | |
AK015990.1 | 3 | 1279 | BAB30069.1 | |
BC113204.1 | 12 | 2278 | AAI13205.1 |