Assay Details
Target Gene Details
Entrez Gene ID: | 70530 |
Gene Name: | leucine rich repeat and fibronectin type III domain containing 2 |
Gene Aliases: |
5730420O05Rik, SALM1, mKIAA1246 |
Location: |
Chr.17:48931566-49097590 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Lrfn2 | NM_027452.3 | 3 | 2327 | NP_081728.2 |
AK017594.1 | 3 | 2531 | BAB30828.1 | |
AK044375.1 | 3 | 2204 | BAC31891.1 | |
AK122476.1 | 3 | 2282 | BAC65758.1 | |
BC116651.1 | ||||
BC139418.1 | 3 | 2327 |
More Information
Set Membership: |
Intragenic Exonic |