Assay Details
Target Gene Details
Entrez Gene ID: | 12648 |
Gene Name: | chromodomain helicase DNA binding protein 1 |
Gene Aliases: |
4930525N21Rik, AI851787, AW555109 |
Location: |
Chr.17:15704967-15772612 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Chd1 | NM_007690.3 | 7 | 1248 | NP_031716.2 |
AK042202.1 | 7 | 1249 | BAC31196.1 | |
AK049686.1 | 7 | 1243 | BAC33872.1 | |
AK140386.1 | 7 | 1228 | BAE24366.1 | |
AK142471.1 | 7 | 1198 | BAE25077.1 | |
BC115822.1 | 6 | 886 | ||
L10410.1 | 7 | 925 | AAB08486.1 |