Assay Details
Target Gene Details
Entrez Gene ID: | 224814 |
Gene Name: | ATP-binding cassette, sub-family C (CFTR/MRP), member 10 |
Gene Aliases: |
Mrp7, mFLJ00002 |
Location: |
Chr.17:46303230-46328032 on Build GRCm38 |
Assay Gene Location: | Within Exon 21 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Abcc10 | NM_145140.2 | 20 | 5386 | NP_660122.1 |
NM_170680.2 | 21 | 4786 | NP_733780.1 | |
AK131108.1 | 18 | 5582 | BAD21358.1 | |
AK133240.1 | 20 | 4818 | BAE21573.1 | |
AK170539.1 | 21 | 4766 | ||
AK171535.1 | 21 | 4765 | ||
BC150818.1 | ||||
CO042416.1 | 1 | 87 |
Target Gene Details
Entrez Gene ID: | 106565 |
Gene Name: | delta-like 2 homolog (Drosophila) |
Gene Aliases: |
AI413481, DLK-2, Egfl9 |
Location: |
Chr.17:46297248-46303271 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Dlk2 | NM_001286013.1 | NP_001272942.1 | ||
NM_001286028.1 | NP_001272957.1 | |||
AB011019.1 | BAA88686.1 | |||
BM935328.1 | ||||
FM180474.1 |
More Information
Set Membership: |
Intragenic Exonic Non-exonic |