Assay Details
Target Gene Details
Entrez Gene ID: | 224814 |
Gene Name: | ATP-binding cassette, sub-family C (CFTR/MRP), member 10 |
Gene Aliases: |
Mrp7, mFLJ00002 |
Location: |
Chr.17:46303230-46328032 on Build GRCm38 |
Assay Gene Location: | Within Exon 17 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Abcc10 | NM_145140.2 | 16 | 4590 | NP_660122.1 |
NM_170680.2 | 17 | 3990 | NP_733780.1 | |
AF406642.1 | 16 | 4590 | AAM18535.1 | |
AF417121.1 | 17 | 3971 | AAM18536.1 | |
AK131108.1 | 14 | 4786 | BAD21358.1 | |
AK133240.1 | 17 | 3947 | BAE21573.1 | |
AK170539.1 | 17 | 3968 | ||
AK170600.1 | 17 | 3964 | BAE41905.1 | |
AK171535.1 | 17 | 3968 | ||
BC150818.1 | 17 | 3943 | ||
CO042416.1 |
More Information
Set Membership: |
Intragenic Exonic |