Assay Details
Target Gene Details
Entrez Gene ID: | 100384868 |
Gene Name: | predicted gene, 37013 |
Gene Aliases: |
Pcdha4-g |
Location: |
Chr.18:36952648-37841873 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gm37013 | NM_001174154.2 | NP_001167625.1 |
Target Gene Details
Entrez Gene ID: | 103611158 |
Gene Name: | predicted gene, 38666 |
Gene Aliases: |
Pcdha6-g |
Location: |
Chr.18:36967631-37841873 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gm38666 | NM_001301256.1 | NP_001288185.1 |
Target Gene Details
Entrez Gene ID: | 103611159 |
Gene Name: | protocadherin alpha 7-gamma |
Gene Aliases: |
- |
Location: |
Chr.18:36973802-37841873 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pcdha7-g | NM_001301259.1 | NP_001288188.1 |
Target Gene Details
Entrez Gene ID: | 93709 |
Gene Name: | protocadherin gamma subfamily A, 1 |
Gene Aliases: |
- |
Location: |
Chr.18:37661945-37841870 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pcdhga1 | NM_033584.1 | NP_291062.1 | ||
AY013792.1 | AAK26081.1 |
Target Gene Details
Entrez Gene ID: | 93722 |
Gene Name: | protocadherin gamma subfamily A, 10 |
Gene Aliases: |
- |
Location: |
Chr.18:37747188-37841870 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pcdhga10 | NM_033593.3 | NP_291071.1 | ||
AY013793.1 | AAK26082.1 | |||
BC035540.1 | AAH35540.1 |
Target Gene Details
Entrez Gene ID: | 93710 |
Gene Name: | protocadherin gamma subfamily A, 2 |
Gene Aliases: |
mKIAA0588 |
Location: |
Chr.18:37669105-37841870 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pcdhga2 | NM_033585.1 | NP_291063.1 | ||
AB093250.1 | BAC41434.1 | |||
AY013796.1 | AAK26085.1 | |||
BC139178.1 |
Target Gene Details
Entrez Gene ID: | 93711 |
Gene Name: | protocadherin gamma subfamily A, 3 |
Gene Aliases: |
- |
Location: |
Chr.18:37674335-37841873 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pcdhga3 | NM_033586.2 | NP_291064.1 | ||
AY013797.1 | AAK26086.1 | |||
BC060691.1 | AAH60691.1 |
Target Gene Details
Entrez Gene ID: | 93712 |
Gene Name: | protocadherin gamma subfamily A, 4 |
Gene Aliases: |
- |
Location: |
Chr.18:37685400-37841870 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pcdhga4 | NM_033587.3 | NP_291065.3 | ||
AY013798.1 | AAK26087.1 | |||
BC049814.1 | AAH49814.1 |
Target Gene Details
Entrez Gene ID: | 93713 |
Gene Name: | protocadherin gamma subfamily A, 5 |
Gene Aliases: |
- |
Location: |
Chr.18:37694501-37841870 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pcdhga5 | NM_033588.4 | NP_291066.1 | ||
AK043491.1 | BAC31558.1 | |||
AY013799.1 | AAK26088.1 | |||
BC057404.1 | AAH57404.1 |
Target Gene Details
Entrez Gene ID: | 93714 |
Gene Name: | protocadherin gamma subfamily A, 6 |
Gene Aliases: |
- |
Location: |
Chr.18:37707229-37841870 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pcdhga6 | NM_033589.1 | NP_291067.1 | ||
AY013800.1 | AAK26089.1 | |||
BC080852.1 | AAH80852.1 |
Target Gene Details
Entrez Gene ID: | 93715 |
Gene Name: | protocadherin gamma subfamily A, 7 |
Gene Aliases: |
- |
Location: |
Chr.18:37714834-37841873 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pcdhga7 | NM_033590.3 | NP_291068.2 | ||
AY013801.1 | AAK26090.1 | |||
BC077712.1 | AAH77712.1 |
Target Gene Details
Entrez Gene ID: | 93716 |
Gene Name: | protocadherin gamma subfamily A, 8 |
Gene Aliases: |
mKIAA4054 |
Location: |
Chr.18:37725706-37841873 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pcdhga8 | NM_033591.3 | NP_291069.1 | ||
AK220461.1 | BAD90279.1 | |||
AY013802.1 | AAK26091.1 | |||
BC054555.1 | AAH54555.1 | |||
BC054787.1 | AAH54787.1 |
Target Gene Details
Entrez Gene ID: | 93717 |
Gene Name: | protocadherin gamma subfamily A, 9 |
Gene Aliases: |
- |
Location: |
Chr.18:37736936-37841863 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pcdhga9 | NM_033592.3 | NP_291070.1 | ||
AY013803.1 | AAK26092.1 | |||
BC079567.1 | AAH79567.1 |
Target Gene Details
Entrez Gene ID: | 93699 |
Gene Name: | protocadherin gamma subfamily B, 1 |
Gene Aliases: |
- |
Location: |
Chr.18:37680458-37841870 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pcdhgb1 | NM_033574.3 | NP_291052.3 | ||
AY013804.1 | AAK26093.1 | |||
BC054741.1 | AAH54741.1 | |||
BC098367.1 | AAH98367.1 |
Target Gene Details
Entrez Gene ID: | 93700 |
Gene Name: | protocadherin gamma subfamily B, 2 |
Gene Aliases: |
- |
Location: |
Chr.18:37689859-37841872 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pcdhgb2 | NM_033575.3 | NP_291053.1 | ||
AF464151.1 | AAM93554.1 | |||
AF464152.1 | AAM93555.1 | |||
AY013805.1 | AAK26094.1 | |||
BC054557.1 | AAH54557.1 |
Target Gene Details
Entrez Gene ID: | 93701 |
Gene Name: | protocadherin gamma subfamily B, 4 |
Gene Aliases: |
- |
Location: |
Chr.18:37720554-37841870 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pcdhgb4 | NM_033576.1 | NP_291054.1 | ||
AY013806.1 | AAK26095.1 | |||
BC138701.1 |
Target Gene Details
Entrez Gene ID: | 93702 |
Gene Name: | protocadherin gamma subfamily B, 5 |
Gene Aliases: |
- |
Location: |
Chr.18:37731154-37841870 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pcdhgb5 | NM_033577.1 | NP_291055.1 | ||
AY013807.1 | AAK26096.1 |
Target Gene Details
Entrez Gene ID: | 93703 |
Gene Name: | protocadherin gamma subfamily B, 6 |
Gene Aliases: |
- |
Location: |
Chr.18:37742094-37841873 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pcdhgb6 | NM_033578.3 | NP_291056.1 | ||
AY013808.1 | AAK26097.1 | |||
BC054556.1 | AAH54556.1 |
Target Gene Details
Entrez Gene ID: | 93704 |
Gene Name: | protocadherin gamma subfamily B, 7 |
Gene Aliases: |
- |
Location: |
Chr.18:37751779-37841870 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pcdhgb7 | NM_033579.1 | 1 | 395 | NP_291057.1 |
AY013809.1 | 1 | 395 | AAK26098.1 | |
BC066823.1 | 1 | 543 | AAH66823.1 |
More Information
Set Membership: |
Intragenic Exonic Intronic Non-exonic |