Assay Details
Target Gene Details
Entrez Gene ID: | 12916 |
Gene Name: | cAMP responsive element modulator |
Gene Aliases: |
ICER, ICERI |
Location: |
Chr.18:3266354-3340355 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Crem | NM_001110851.1 | NP_001104321.1 | ||
NM_001110852.1 | NP_001104322.1 | |||
AK010806.1 | BAB27192.1 |
Target Gene Details
Entrez Gene ID: | 633947 |
Gene Name: | predicted gene 6225 |
Gene Aliases: |
EG621440 |
Location: |
Chr.18:3336416-3366863 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gm6225 | NR_033457.1 | |||
AK086784.1 | ||||
AK157308.1 | 1 | 852 | ||
AK163460.1 | ||||
AK166419.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic |