Assay Details
Target Gene Details
Entrez Gene ID: | 53901 |
Gene Name: | regulator of calcineurin 2 |
Gene Aliases: |
Csp2, Dscr1l1, MCIP2, ZAKI-4 |
Location: |
Chr.17:43801281-44039516 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Rcan2 | NM_001286653.1 | 5 | 1960 | NP_001273582.1 |
NM_001286654.1 | 5 | 1837 | NP_001273583.1 | |
NM_030598.2 | 4 | 1967 | NP_085101.1 | |
NM_207649.1 | 5 | 2093 | NP_997532.1 | |
AB061524.1 | 4 | 1905 | BAB71955.1 | |
AB061525.1 | 5 | 1960 | BAB71956.1 | |
AK049006.1 | 4 | 1906 | BAC33511.1 | |
AK136805.1 | 5 | 1902 | ||
AK138768.1 | 5 | 2005 | BAE23773.1 | |
AK160865.1 | 4 | 1855 | BAE36057.1 | |
BC047153.1 | 4 | 1765 | AAH47153.1 | |
BC049096.1 | 4 | 1967 | AAH49096.1 |
More Information
Set Membership: |
Intragenic Exonic |