Assay Details
Target Gene Details
Entrez Gene ID: | 14670 |
Gene Name: | guanine nucleotide binding protein-like 1 |
Gene Aliases: |
Gna-rs1, Gnal1 |
Location: |
Chr.17:35979955-35989462 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gnl1 | NM_008136.2 | 12 | 2433 | NP_032162.2 |
AK078187.1 | 12 | 2396 | ||
AK210008.1 | 1 | 174 | ||
AV338067.2 | 1 | 351 | ||
BC094391.1 | 10 | 1870 | AAH94391.1 |