Assay Details
Target Gene Details
Entrez Gene ID: | 224419 |
Gene Name: | Map3k7 C-terminal like |
Gene Aliases: |
C21orf7, ORF63, Tak1l |
Location: |
Chr.16:87553330-87595336 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Map3k7cl | NM_144854.2 | 1 | 86 | NP_659103.1 |
AK041664.1 | BAC31024.1 | |||
AY033899.1 | 1 | 101 | AAK68717.1 | |
BX519631.1 | 1 | 87 |