Assay Details
Target Gene Details
Entrez Gene ID: | 78923 |
Gene Name: | chondroitin sulfate synthase 3 |
Gene Aliases: |
4833446K15Rik, AI662215 |
Location: |
Chr.18:59175340-59411336 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Chsy3 | NM_001081328.1 | 3 | 2650 | NP_001074797.1 |
AK019528.1 | 3 | 1637 | BAC25596.1 | |
BC151164.1 | 3 | 2340 | ||
BC151165.1 | 3 | 2340 | ||
BC157985.1 | 3 | 2340 |
More Information
Set Membership: |
Intragenic Exonic |