Assay Details
Target Gene Details
Entrez Gene ID: | 26369 |
Gene Name: | centrin 1 |
Gene Aliases: |
caltractin |
Location: |
Chr.18:9618419-9619469 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cetn1 | NM_007593.5 | 1 | 351 | NP_031619.3 |
AK005736.1 | 1 | 330 | BAB24213.1 | |
AK005743.1 | 1 | 312 | BAB24217.1 | |
AK005837.1 | 1 | 352 | BAB24266.1 | |
AK006940.1 | 1 | 352 | BAB24798.1 | |
AK132753.1 | 1 | 351 | BAE21334.1 | |
BC048488.1 | 1 | 352 | AAH48488.1 | |
BC061155.1 | 1 | 350 | AAH61155.1 | |
BU961997.1 | 1 | 389 | ||
D16301.1 | 1 | 335 | BAA03806.1 |
More Information
Set Membership: |
Intragenic Exonic |