Assay Details
Target Gene Details
Entrez Gene ID: | 52563 |
Gene Name: | CDC23 cell division cycle 23 |
Gene Aliases: |
6030435O18, D18Ertd243e |
Location: |
Chr.18:34630946-34651745 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cdc23 | NM_178347.4 | 16 | 3301 | NP_848124.1 |
AK047789.1 | ||||
AK052567.1 | ||||
AK075948.1 | ||||
AK081461.1 | 16 | 3302 | BAC38224.1 |
Target Gene Details
Entrez Gene ID: | 19348 |
Gene Name: | kinesin family member 20A |
Gene Aliases: |
AA415432, Rab6kifl |
Location: |
Chr.18:34624624-34633277 on Build GRCm38 |
Assay Gene Location: | Within Exon 20 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Kif20a | NM_001166406.1 | 19 | 2818 | NP_001159878.1 |
NM_001166407.1 | 19 | 2816 | NP_001159879.1 | |
NM_009004.4 | 19 | 2712 | NP_033030.1 | |
AK051201.1 | 19 | 2631 | ||
AK051995.1 | 19 | 2667 | ||
AK053280.1 | 19 | 2631 | ||
AK083412.1 | 19 | 2672 | BAC38906.1 | |
AK083697.1 | 19 | 2633 | ||
AK084732.1 | 19 | 2654 | BAC39267.1 | |
AK142764.1 | 19 | 2660 | ||
AK144844.1 | 19 | 2630 | BAE26095.1 | |
BC060608.1 | 19 | 2818 | AAH60608.1 | |
Y09632.1 | 19 | 2867 | CAA70845.1 |
More Information
Set Membership: |
Intragenic Exonic |