Assay Details
Target Gene Details
Entrez Gene ID: | 12393 |
Gene Name: | runt related transcription factor 2 |
Gene Aliases: |
AML3, Cbf, Cbfa-1, Cbfa1, LS3, Osf2, PEBP2aA, Pebp2a1, Pebpa2a |
Location: |
Chr.17:44495986-44814831 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Runx2 | NM_001146038.2 | NP_001139510.1 | ||
NM_001271627.1 | NP_001258556.1 | |||
NM_009820.5 | NP_033950.2 | |||
NR_073392.1 | ||||
AF005936.1 | AAB82419.1 | |||
AF010284.1 | AAB65409.1 | |||
AF053954.1 | ||||
AF053955.1 | ||||
DQ458792.1 | ABE68713.1 |
Target Gene Details
Entrez Gene ID: | 109115 |
Gene Name: | suppressor of Ty 3 |
Gene Aliases: |
2310066G22Rik, AI315192, SPT3, SPT3L, Supt3h |
Location: |
Chr.17:44777171-45119294 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Supt3 | NM_178652.2 | 1 | 89 | NP_848767.1 |
AK075916.1 | 1 | 89 | BAC36052.1 |
More Information
Set Membership: |
Intragenic Exonic Intronic Non-exonic |