Assay Details
Target Gene Details
Entrez Gene ID: | 74556 |
Gene Name: | thymocyte selection associated family member 3 |
Gene Aliases: |
9130404H23Rik |
Location: |
Chr.17:66555252-66594622 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Themis3 | NM_028998.1 | 4 | 1633 | NP_083274.1 |
AK018663.1 | 4 | 1634 | BAB31332.1 |