Assay Details
Target Gene Details
Entrez Gene ID: | 171286 |
Gene Name: | solute carrier family 12 (potassium/chloride transporters), member 8 |
Gene Aliases: |
CCC9, E330020C02Rik |
Location: |
Chr.16:33517333-33664135 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Slc12a8 | NM_001083902.1 | 12 | 2640 | NP_001077371.1 |
NM_134251.2 | 13 | 2756 | NP_599012.2 | |
AK165080.1 | 8 | 2263 | BAE38029.1 |
More Information
Set Membership: |
Intragenic Exonic |