Assay Details
Target Gene Details
Entrez Gene ID: | 22661 |
Gene Name: | zinc finger protein 148 |
Gene Aliases: |
2210405J08Rik, AI480666, AW045217, BERF-1, BFCOL1, ZBP-89, Znf148 |
Location: |
Chr.16:33380775-33503903 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Zfp148 | NM_011749.4 | 1 | 216 | NP_035879.1 |
AF316548.1 | 1 | 155 | AAK63003.1 | |
BC026144.1 | AAH26144.1 | |||
CJ129103.1 | 1 | 217 | ||
X98096.1 | 1 | 113 | CAA66725.1 |
More Information
Set Membership: |
Intragenic Exonic |