Assay Details
Target Gene Details
Entrez Gene ID: | 224432 |
Gene Name: | SR-related CTD-associated factor 4 |
Gene Aliases: |
AA517739, Sfrs15, Sra4, Srsf15 |
Location: |
Chr.16:90228951-90284498 on Build GRCm38 |
Assay Gene Location: | Within Exon 20 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Scaf4 | NM_178923.3 | 20 | 3631 | NP_849254.2 |
AK045266.1 | 20 | 3619 | ||
AK173120.1 | 20 | 3634 | BAD32398.1 | |
BC024860.1 | 2 | 759 | AAH24860.1 | |
BC053096.1 | 20 | 3448 | AAH53096.1 | |
BC057592.1 | 20 | 3631 | AAH57592.1 |
More Information
Set Membership: |
Intragenic Exonic |