Assay Details
Target Gene Details
Entrez Gene ID: | 19252 |
Gene Name: | dual specificity phosphatase 1 |
Gene Aliases: |
3CH134, MKP1, Ptpn16, U19515, erp, mkp-1 |
Location: |
Chr.17:26505591-26508472 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Dusp1 | NM_013642.3 | 1 | 86 | NP_038670.1 |
AK145903.1 | 1 | 89 | ||
AK149915.1 | BAE29163.1 | |||
AK150433.1 | 1 | 86 | BAE29556.1 | |
AK152183.1 | 1 | 86 | BAE31014.1 | |
AK152324.1 | 1 | 89 | BAE31126.1 | |
AK152905.1 | 1 | 87 | ||
AK159251.1 | 1 | 87 | BAE34932.1 | |
AK159363.1 | 1 | 87 | BAE35021.1 | |
AK159567.1 | 1 | 87 | BAE35190.1 | |
AK159658.1 | 1 | 84 | BAE35267.1 | |
AK159726.1 | BAE35320.1 | |||
AK167602.1 | 1 | 88 | BAE39659.1 | |
AK168627.1 | 1 | 87 | BAE40487.1 | |
BC006967.1 | 1 | 73 | AAH06967.1 | |
X61940.1 | 1 | 76 | CAA43944.1 |
More Information
Set Membership: |
Intragenic Exonic |